Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003449.1 | NP_001003449.1 | 5 - 6 | 385 | 70 | |
GenBank mRNA | BC076054.1 | - | 5 - 6 | 385 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |