Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc85al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004570.1 | NP_001004570.1 | 2 - 3 | 525 | 133 | |
XM_005165709.4 | XP_005165766.1 | 1 - 2 | 266 | 133 | ||
XM_005165710.4 | XP_005165767.1 | 1 - 2 | 266 | 133 | ||
XM_009302126.3 | XP_009300401.1 | 1 - 2 | 266 | 133 | ||
XM_009302127.3 | XP_009300402.1 | 1 - 2 | 266 | 133 | ||
GenBank mRNA | BC081674.1 | - | 2 - 3 | 525 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |