Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alkbh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005390.1 | NP_001005390.1 | 5 - 6 | 620 | 103 | |
GenBank mRNA | BC071457.1 | - | 5 - 6 | 620 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |