Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctdnep1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007309.1 | NP_001007310.1 | 4 - 5 | 863 | 99 | |
XM_021476127.1 | XP_021331802.1 | 5 - 6 | 765 | 99 | ||
GenBank mRNA | BC085649.1 | - | 4 - 5 | 863 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |