Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hif1al2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012371.2 | NP_001012371.2 | 1 - 2 | 178 | 75 | |
XM_005161350.4 | XP_005161407.1 | 2 - 3 | 515 | 75 | ||
XM_021468817.1 | XP_021324492.1 | 3 - 4 | 391 | 75 | ||
GenBank mRNA | AY835381.1 | - | 1 - 2 | 91 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Danio rerio Probe spans exons |