Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crygs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013275.1 | NP_001013293.1 | 2 - 3 | 259 | 85 | |
GenBank mRNA | AY738753.1 | - | 2 - 3 | 259 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |