Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: myoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015062.2 | NP_001015062.1 | 1 - 2 | 777 | 77 | |
GenBank mRNA | AY551079.1 | - | 1 - 2 | 475 | 77 | |
BC115210.1 | - | 1 - 2 | 777 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |