Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrna5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017885.1 | NP_001017885.1 | 2 - 3 | 177 | 81 | |
GenBank mRNA | BC092962.1 | - | 2 - 3 | 177 | 81 | |
BC164212.1 | - | 1 - 2 | 104 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |