Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atpaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017907.2 | NP_001017907.2 | 2 - 3 | 322 | 139 | |
XM_021469326.1 | XP_021325001.1 | 2 - 3 | 247 | 139 | ||
GenBank mRNA | BC092940.1 | - | 2 - 3 | 323 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |