Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020532.1 | NP_001018368.1 | 4 - 5 | 635 | 82 | |
GenBank mRNA | BC078380.1 | - | 4 - 5 | 621 | 82 | |
BC095067.1 | - | 4 - 5 | 635 | 82 | ||
BC152166.1 | - | 4 - 5 | 635 | 82 | ||
BC165441.1 | - | 4 - 5 | 640 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |