Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lactb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020593.1 | NP_001018429.1 | 5 - 6 | 1205 | 77 | |
XM_009298158.3 | XP_009296433.1 | 5 - 6 | 1213 | 77 | ||
GenBank mRNA | BC095252.1 | - | 5 - 6 | 1205 | 77 | |
BC165373.1 | - | 5 - 6 | 1114 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |