Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020733.2 | NP_001018569.2 | 2 - 3 | 319 | 88 | |
GenBank mRNA | AY584746.1 | - | 2 - 3 | 319 | 88 | |
BC095116.1 | - | 2 - 3 | 351 | 88 | ||
BC165891.1 | - | 2 - 3 | 185 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |