Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daam1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030136.2 | NP_001025307.1 | 22 - 23 | 2722 | 61 | |
XM_005160502.4 | XP_005160559.1 | 23 - 24 | 2929 | 61 | ||
XM_005160503.4 | XP_005160560.1 | 23 - 24 | 3123 | 61 | ||
XM_017352491.2 | XP_017207980.1 | 22 - 23 | 3093 | 61 | ||
GenBank mRNA | BC163491.1 | - | 21 - 22 | 2725 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |