Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033731.1 | NP_001028903.1 | 3 - 4 | 421 | 100 | |
GenBank mRNA | BC097192.1 | - | 3 - 4 | 421 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |