Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044832.3 | NP_001038297.3 | 5 - 6 | 712 | 69 | |
NM_001328060.1 | NP_001314989.1 | 5 - 6 | 1683 | 69 | ||
GenBank mRNA | BC134855.1 | - | 5 - 6 | 712 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |