Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lcn15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045415.1 | NP_001038880.1 | 1 - 2 | 143 | 59 | |
XM_005172030.4 | XP_005172087.2 | 1 - 2 | 277 | 59 | ||
GenBank mRNA | BC122437.1 | - | 1 - 2 | 143 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |