Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lcn15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045415.1 | NP_001038880.1 | 6 - 7 | 597 | 93 | |
GenBank mRNA | BC122437.1 | - | 6 - 7 | 597 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |