Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045852.1 | NP_001039317.1 | 3 - 4 | 539 | 101 | |
XM_005165334.2 | XP_005165391.1 | 3 - 4 | 694 | 101 | ||
GenBank mRNA | BC122226.1 | - | 3 - 4 | 539 | 101 | |
BC164250.1 | - | 3 - 4 | 370 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |