Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: desmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077452.1 | NP_001070920.1 | 8 - 9 | 1424 | 59 | |
GenBank mRNA | BC124512.1 | - | 8 - 9 | 1424 | 59 | |
BC165598.1 | - | 8 - 9 | 1419 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |