Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ino80db | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080588.1 | NP_001074057.1 | 3 - 4 | 1380 | 128 | |
GenBank mRNA | BC129037.1 | - | 3 - 4 | 1380 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |