Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clcn1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082910.1 | NP_001076379.1 | 22 - 23 | 2450 | 141 | |
XM_009294060.3 | XP_009292335.1 | 22 - 23 | 2781 | 141 | ||
XM_017352282.2 | XP_017207771.1 | 21 - 22 | 2652 | 141 | ||
XM_017352285.2 | XP_017207774.1 | 14 - 15 | 1557 | 141 | ||
XM_021468144.1 | XP_021323819.1 | 22 - 23 | 2553 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Danio rerio Probe spans exons |