Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc22a7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083861.1 | NP_001077330.1 | 4 - 5 | 938 | 63 | |
GenBank mRNA | BC134964.1 | - | 4 - 5 | 938 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |