Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npffr1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089389.1 | NP_001082858.1 | 2 - 3 | 475 | 83 | |
GenBank mRNA | BC134148.1 | - | 2 - 3 | 475 | 83 | |
GU290221.1 | - | 2 - 3 | 492 | 83 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |