Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc25a35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099447.1 | NP_001092917.1 | 3 - 4 | 592 | 72 | |
XM_005165493.4 | XP_005165550.1 | 3 - 4 | 1082 | 72 | ||
GenBank mRNA | BC171554.1 | - | 3 - 4 | 670 | 72 | |
BC171721.1 | - | 3 - 4 | 669 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |