Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100012.2 | NP_001093482.1 | 7 - 8 | 1015 | 125 | |
XM_005159014.3 | XP_005159071.1 | 7 - 8 | 1284 | 125 | ||
GenBank mRNA | BC163504.1 | - | 7 - 8 | 1176 | 125 | |
BC163518.1 | - | 7 - 8 | 1176 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |