Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109825.1 | NP_001103295.1 | 2 - 3 | 331 | 64 | |
GenBank mRNA | DQ986317.1 | - | 2 - 3 | 331 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |