Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131005.2 | NP_571080.1 | 2 - 3 | 160 | 65 | |
GenBank mRNA | BC092712.1 | - | 2 - 3 | 160 | 65 | |
X52502.1 | - | 130 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |