Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131009.1 | NP_571084.1 | 6 - 7 | 863 | 77 | |
GenBank mRNA | BC045289.1 | - | 6 - 7 | 866 | 77 | |
BC164456.1 | - | 6 - 7 | 604 | 77 | ||
U70479.1 | - | 6 - 7 | 863 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |