Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppm1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131398.1 | NP_571473.1 | 2 - 3 | 902 | 115 | |
GenBank mRNA | BC079530.1 | - | 2 - 3 | 902 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |