Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppm1ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131398.1 | NP_571473.1 | 3 - 4 | 1013 | 71 | |
GenBank mRNA | BC079530.1 | - | 3 - 4 | 1013 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |