Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: krt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131509.2 | NP_571584.2 | 7 - 8 | 1377 | 82 | |
GenBank mRNA | AF134850.1 | - | 7 - 8 | 1361 | 82 | |
BC066728.1 | - | 7 - 8 | 1366 | 82 | ||
BC155774.1 | - | 7 - 8 | 1377 | 82 | ||
EH999878.1 | - | 4 - 5 | 606 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |