Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scn8aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131628.1 | NP_571703.1 | 23 - 24 | 4474 | 135 | |
XM_005162268.4 | XP_005162325.1 | 23 - 24 | 4518 | 135 | ||
XM_009296871.3 | XP_009295146.1 | 23 - 24 | 4518 | 135 | ||
XM_009296874.3 | XP_009295149.1 | 23 - 24 | 4484 | 135 | ||
XM_021469841.1 | XP_021325516.1 | 23 - 24 | 4663 | 135 | ||
GenBank mRNA | AF297658.1 | - | 23 - 24 | 4474 | 135 | |
BC163773.1 | - | 23 - 24 | 4326 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |