Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183349.1 | NP_899192.1 | 15 - 16 | 2493 | 59 | |
GenBank mRNA | AY313897.1 | - | 15 - 16 | 2493 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |