Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200022.1 | NP_956316.1 | 9 - 10 | 786 | 86 | |
GenBank mRNA | BC057434.1 | - | 9 - 10 | 786 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |