Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snapc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200263.1 | NP_956557.1 | 10 - 11 | 1065 | 67 | |
XM_021468375.1 | XP_021324050.1 | 8 - 9 | 67 | |||
XM_021468376.1 | XP_021324051.1 | 9 - 10 | 67 | |||
GenBank mRNA | BC049022.1 | - | 10 - 11 | 1065 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |