Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200713.1 | NP_957007.1 | 4 - 5 | 624 | 123 | |
XM_005167684.1 | XP_005167741.1 | 5 - 6 | 1195 | 123 | ||
GenBank mRNA | BC059473.1 | - | 4 - 5 | 624 | 123 | |
BC065432.1 | - | 4 - 5 | 621 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |