Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpe65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200751.1 | NP_957045.1 | 9 - 10 | 1027 | 67 | |
GenBank mRNA | AY646886.1 | - | 8 - 9 | 994 | 67 | |
BC059559.1 | - | 9 - 10 | 1027 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |