Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nucb2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201493.1 | NP_958901.1 | 7 - 8 | 871 | 112 | |
GenBank mRNA | BC046077.1 | - | 7 - 8 | 871 | 112 | |
BC065437.1 | - | 7 - 8 | 837 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |