Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult1st4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205620.2 | NP_991183.1 | 6 - 7 | 672 | 84 | |
XM_021478222.1 | XP_021333897.1 | 6 - 7 | 842 | 84 | ||
XM_021478223.1 | XP_021333898.1 | 6 - 7 | 1155 | 84 | ||
XM_021478224.1 | XP_021333899.1 | 6 - 7 | 832 | 84 | ||
GenBank mRNA | AY196986.1 | - | 6 - 7 | 629 | 84 | |
BC066584.1 | - | 6 - 7 | 672 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |