Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205746.1 | NP_991309.1 | 3 - 4 | 450 | 63 | |
XM_021476820.1 | XP_021332495.1 | 1 - 2 | 143 | 64 | ||
GenBank mRNA | AY394949.1 | - | 3 - 4 | 450 | 63 | |
BC075882.1 | - | 3 - 4 | 437 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |