Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205746.1 | NP_991309.1 | 4 - 5 | 654 | 75 | |
XM_021476820.1 | XP_021332495.1 | 2 - 3 | 347 | 75 | ||
GenBank mRNA | AY394949.1 | - | 4 - 5 | 654 | 75 | |
BC075882.1 | - | 4 - 5 | 641 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |