Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dip2ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212963.1 | NP_998128.1 | 26 - 27 | 3315 | 75 | |
XM_009296840.3 | XP_009295115.1 | 28 - 29 | 3383 | 75 | ||
XM_009296841.3 | XP_009295116.1 | 27 - 28 | 3377 | 75 | ||
XM_009296842.3 | XP_009295117.1 | 27 - 28 | 3359 | 75 | ||
GenBank mRNA | BC068015.1 | - | 26 - 27 | 3315 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |