Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubqln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213356.2 | NP_998521.2 | 10 - 11 | 1663 | 70 | |
GenBank mRNA | BC055536.1 | - | 8 - 9 | 1365 | 70 | |
BC098879.1 | - | 10 - 11 | 1663 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |