Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001310068.1 | NP_001296997.1 | 5 - 6 | 861 | 95 | |
NM_214795.2 | NP_999960.2 | 5 - 6 | 853 | 95 | ||
GenBank mRNA | AB125219.1 | - | 5 - 6 | 853 | 95 | |
AY616134.1 | - | 5 - 6 | 647 | 95 | ||
BC162621.1 | - | 5 - 6 | 611 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |