Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002145.1 | NP_001002145.1 | 6 - 7 | 823 | 101 | |
GenBank mRNA | BC071393.1 | - | 6 - 7 | 823 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |