Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: leng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004591.1 | NP_001004591.1 | 2 - 3 | 613 | 110 | |
GenBank mRNA | BC081609.1 | - | 2 - 3 | 613 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |