Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xgb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012261.2 | NP_001012261.2 | 4 - 5 | 554 | 78 | |
XM_021467154.1 | XP_021322829.1 | 3 - 4 | 604 | 78 | ||
GenBank mRNA | BC133862.1 | - | 4 - 5 | 554 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |