Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: shisa9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013509.1 | NP_001013527.1 | 1 - 2 | 783 | 69 | |
GenBank mRNA | BC090494.1 | - | 1 - 2 | 783 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |