Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtf2f2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017832.1 | NP_001017832.1 | 7 - 8 | 723 | 94 | |
GenBank mRNA | BC092783.1 | - | 7 - 8 | 723 | 94 | |
BC165452.1 | - | 7 - 8 | 661 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |