Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtf2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020605.1 | NP_001018441.1 | 1 - 2 | 177 | 143 | |
GenBank mRNA | BC095322.1 | - | 1 - 2 | 177 | 143 | |
BC165482.1 | - | 1 - 2 | 108 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |